missing translation for 'onlineSavingsMsg'
Learn More
Learn More
T-box 19 Antibody (CL6251) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61438JF525
This item is not returnable.
View return policy
Description
T-box 19 Monoclonal antibody specifically detects T-box 19 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| T-box 19 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| dJ747L4.1, FLJ26302, FLJ34085, T-box 19, T-box factor, pituitary, T-box protein 19, T-box transcription factor TBX19, TBS 19, TBS19, TPITFLJ34543 | |
| This antibody was developed using a recombinant protein derived from O60806, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL6251 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 9095 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction