Learn More
T-box 19 Antibody (CL6251) [PerCP], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-61438PCP
Product Code. 30301768
  • 737.00 € / 0.10mL
Description
T-box 19 Monoclonal antibody specifically detects T-box 19 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
T-box 19
Monoclonal
PerCP
dJ747L4.1, FLJ26302, FLJ34085, T-box 19, T-box factor, pituitary, T-box protein 19, T-box transcription factor TBX19, TBS 19, TBS19, TPITFLJ34543
This antibody was developed using a recombinant protein derived from O60806, with the exact immunogen sequence remaining proprietary.
0.1 mL
Primary
Human
Purified
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL6251
PBS
Mouse
Protein A purified
RUO
9095
Store at 4⪚C in the dark.
IgG1