missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TAAR2 Polyclonal Antibody

Description
Antibody detects endogenous levels of total TAAR2.
Orphan receptor. [UniProt].
Specifications
Specifications
| Antigen | TAAR2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | TAAR2 |
| Gene Accession No. | Q5QD17, Q5QD25, Q9P1P5 |
| Gene Alias | G protein-coupled receptor 58; Gm224; Gm225; GPR58; G-protein coupled receptor 58; mTaar2; PhBL5; TAAR2; taR-2; Tarr2; trace amine associated receptor 2; trace amine receptor 2; trace amine-associated receptor 2 |
| Gene Symbols | TAAR2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?