missing translation for 'onlineSavingsMsg'
Learn More

TAAR3 Polyclonal Antibody, Invitrogen™

Product Code. 15722546
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15722546 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15722546 Supplier Invitrogen Supplier No. PA3047

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IHC(P) analysis shows positive staining of GPR57 in ascending tubules of human kidney. WB analysis shows GPR57 in glyco-protein enriched fractions from mock transfected or GPR57 overexpressed 293 cells. GPR57 exists in different glycosylated forms.

G protein-coupled receptors (GPCRs, or GPRs) contain 7 transmembrane domains and transduce extracellular signals through heterotrimeric G proteins. Expression of GPR101 has been documented by Northern analysis in human hypothalamus, caudate, and putamen but not in thalamus, hippocampus, frontal cortex, or pons.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAAR3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation whole serum with 0.05% sodium azide
Gene TAAR3P
Gene Accession No. Q9P1P4
Gene Alias GPR57, GPR58, TAAR3, G protein-coupled receptor 57, Putative trace amine-associated receptor 3, TAAR2
Gene Symbols TAAR3P
Host Species Rabbit
Immunogen KLH conjugated COOH-terminal amino acids of 324-343 (VSGKIFSSHSETANLFPEAH)
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9288
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.