Learn More
Invitrogen™ TAB3 Recombinant Rabbit Monoclonal Antibody (24GB2680)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557278
 View more versions of this product
Product Code. 30276119
  • 146.00 € / 20µL
Description
TAB3 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
TAB3 functions in the NF-kappaB signal transduction pathway. It and the similar and functionally redundant protein TAB2, form a ternary complex with the protein kinase TAK1 and either TRAF2 or TRAF6 in response to stimulation with the pro-inflammatory cytokines TNF or IL-1. Subsequent TAK1 kinase activity triggers a signaling cascade leading to activation of the NF-kappaB transcription factor. Recent experiments have shown that TAB2 and the related protein TAB3 constitutitvely interact with the autophagy mediator Beclin-1; upon induction of autophagy, these proteins dissociate from Beclin-1 and bind TAK1. Overexpression of TAB2 and TAB3 inhibit autophagy, while their depletion triggers it, suggesting that TAB2 and TAB3 act as a control point for autophagy.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TAB3
Recombinant Monoclonal
Unconjugated
Tab3
4921526G09Rik; Kiaa4135; MAP3K7IP3; mitogen activated protein kinase kinase kinase 7 interacting protein 3; mitogen-activated protein kinase kinase kinase 7 interacting protein 3; mitogen-activated protein kinase kinase kinase 7-interacting protein 3; mKIAA4135; NAP1; NF-kappa-B-activating protein 1; NFkB activating protein 1; Tab3; TAB-3; TAK1-binding protein 3; TGF-beta activated kinase 1 (MAP3K7) binding protein 3; TGF-beta activated kinase 1 and MAP3K7 binding protein 3; TGF-beta activated kinase 1/MAP3K7 binding protein 3; TGF-beta-activated kinase 1 and MAP3K7-binding protein 3; TGF-beta-activated kinase 1-binding protein 3
Rabbit
Affinity chromatography
RUO
257397, 66724
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB2680
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q571K4, Q8N5C8
Tab3
A synthesized peptide derived from human TAB3 (aa 50-94).
20 μL
Primary
Human, Mouse