Learn More
Invitrogen™ TACC3 Recombinant Rabbit Monoclonal Antibody (24GB1940)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557756
 View more versions of this product
Product Code. 30276169
  • 564.00 € / 100µL
Description
TACC3 Recombinant Monoclonal Antibody for Western Blot, Flow
TACC1 is located on 8p11 chromosomal region that is amplified in approximately 15% of all breast tumor samples. The short arm of chromosome 8 also contains FGFR1 whose expression is enhanced in most breast cancer tumors. TACC family members, TACC1, TACC2, and TACC3, map very closely to the corresponding FGFR1, FGFR2, FGFR3 genes on chromosomes 4,8, and 10. Subsequently, since they are phylogenetically related, it is proposed that TACC and FGFR have similar roles in cell growth and differentiation. Also, TACC1 contains a conserved C-terminal region as in the Drosophila homolog, D-TACC. It has been shown that D-TACC is necessary for normal spindle function, and the mammalian TACC proteins appears to interact with centrosomes and microtubules in a similar manner.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TACC3
Recombinant Monoclonal
Unconjugated
TACC3
Aint; Arnt interacting protein; ARNT-interacting protein; C86661; ERIC1; ERIC-1; Tacc3; transforming acidic coiled coil 3; transforming acidic coiled-coil containing protein 3; transforming acidic coiled-coil-containing protein 3; transforming, acidic coiled-coil containing protein 3
Rabbit
Affinity chromatography
RUO
10460, 360962
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB1940
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q9Y6A5
TACC3
A synthesized peptide derived from human TACC3 (aa 50-100).
100 μL
Primary
Human, Rat