missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TACC3 Recombinant Rabbit Monoclonal Antibody (24GB1940)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557757
This item is not returnable.
View return policy
Description
TACC3 Recombinant Monoclonal Antibody for Western Blot, Flow
TACC1 is located on 8p11 chromosomal region that is amplified in approximately 15% of all breast tumor samples. The short arm of chromosome 8 also contains FGFR1 whose expression is enhanced in most breast cancer tumors. TACC family members, TACC1, TACC2, and TACC3, map very closely to the corresponding FGFR1, FGFR2, FGFR3 genes on chromosomes 4,8, and 10. Subsequently, since they are phylogenetically related, it is proposed that TACC and FGFR have similar roles in cell growth and differentiation. Also, TACC1 contains a conserved C-terminal region as in the Drosophila homolog, D-TACC. It has been shown that D-TACC is necessary for normal spindle function, and the mammalian TACC proteins appears to interact with centrosomes and microtubules in a similar manner.
Specifications
| TACC3 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| TACC3 | |
| Aint; Arnt interacting protein; ARNT-interacting protein; C86661; ERIC1; ERIC-1; Tacc3; transforming acidic coiled coil 3; transforming acidic coiled-coil containing protein 3; transforming acidic coiled-coil-containing protein 3; transforming, acidic coiled-coil containing protein 3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10460, 360962 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB1940 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9Y6A5 | |
| TACC3 | |
| A synthesized peptide derived from human TACC3 (aa 50-100). | |
| 20 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction