Learn More
Invitrogen™ Tadocizumab Recombinant Monoclonal Antibody
GREENER_CHOICE
Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA559797
 View more versions of this product
Product Code. 30284305
  • 477.00 € / 100µg
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Tadocizumab is a biosimilar that targets ITGA2B & ITGB3. CD41 is a transmembrane glycoprotein (integrin family) composed of two chains GpiIb alpha (heavy chain; 120 kDa) and GpiIb beta (light chain; 23 kDa). CD61 non-covalently associates with CD41.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Tadocizumab Humanized
Recombinant Monoclonal
Unconjugated
C4G1; YM-337
Protein A
RUO
Human
Antibody
IgG1
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Human
100 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized