Learn More
Thermo Scientific Chemicals TAE (50X), TRIS + acetate + EDTA
TAE (50X), TRIS + acetate + EDTA is a buffer composed of Tris, acetate, and EDTA components and is used in electrophoresis of nucleic acids in agarose and polyacrylamide gels.
Brand: Thermo Scientific Chemicals J63931.K7
Code : NEW
Additional Details : Weight : 4.00000kg
Description
TAE (50X), TRIS + acetate + EDTA is used in electrophoresis of nucleic acids in agarose and polyacrylamide gel. It acts as a running buffer and as a gel preparation buffer. It is also useful for nucleic acid separations viz. DNA and RNA. Further, it is used for non-denaturing RNA agarose gel electrophoresis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• TAE (50X), TRIS + acetate + EDTA allows nucleic acid separation
Application
• This buffer has been used to prepare agarose and polyacrylamide gels for electrophoresis of nucleic acids
• It is used as a running and gel preparation buffer, and it is also used for non-denaturing RNA agarose gel electrophoresis
Specifications
Buffer | |
50X | |
4 L |
Colorless | |
Liquid |
Safety and Handling
- TAE (50X), TRIS + acetate + EDTA
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Acetic acid
RUO – Research Use Only