missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAE Buffer, 10X Solution, Molecular Biology Grade, Ultrapure
Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 L
5 L
Packaging:
Nalgene bottle
Upright carton
Unit Size:
1 L
5 L
Spécification
Spécification
| Chemical Name or Material | TAE Buffer |
| Concentration | 10X |
| Physical Form | Solution |
| Quantity | 1 L |
| Packaging | Nalgene bottle |
| Grade | Molecular Biology, Ultrapure |
Frequently Asked Questions (FAQs)
What are the concentrations of the components of TAE Buffer, 10X Solution, Molecular Biology Grade, Ultrapure (Cat. No. J75904.K8)?
TAE Buffer, 10X Solution, Molecular Biology Grade, Ultrapure contains 400 mM Tris and 0.01 M EDTA in high-purity RO water.
RUO – Research Use Only
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu