missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAF1B Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93179-0.02ml
This item is not returnable.
View return policy
Description
TAF1B Polyclonal antibody specifically detects TAF1B in Human, Mouse samples. It is validated for Western Blot
Specifications
| TAF1B | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| RAF1BTATA box binding protein (TBP)-associated factor, RNA polymerase I, B, 63kD, RAFI63MGC:9349, RNA polymerase I-specific TBP-associated factor 63 kDa, SL1SL1, 63kD subunit, TAFI63TBP-associated factor 1B, TATA box binding protein (TBP)-associated factor, RNA polymerase I, B, 63kDa, TATA box-binding protein-associated factor 1B, TATA box-binding protein-associated factor RNA polymerase I subunit B, Transcription initiation factor SL1/TIF-IB subunit B | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 369-588 of human TAF1B (NP_005671.2). LFLLDDSFEWSLSNLAEKHNEKNKKDKPWFDFRKWYQIMKKAFDEKKQKWEEARAKYLWKSEKPLYYSFVDKPVAYKKREMVVNLQKQFSTLVESTATAGKKSPSSFQFNWTEEDTDRTCFHGHSLQGVLKEKGQSLLTKNSLYWLSTQKFCRCYCTHVTTYEESNYSLSYQFILNLFSFLLRIKTSLLHEEVSLVEKKLFEKKYSVKRKKSRSKKVRRH | |
| 0.02 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 9014 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction