missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAF1L Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-30752
This item is not returnable.
View return policy
Description
TAF1L Polyclonal specifically detects TAF1L in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| TAF1L | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| Q8IZX4 | |
| TAF1L | |
| This antibody was developed against a recombinant protein corresponding to amino acids: PKQPFMLQHASGEHKDGHGK | |
| 0.1 mL | |
| Protein Kinase | |
| 138474 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 210kDa, 210kDa-like, EC 2.7.11, EC 2.7.11.1, MGC134910, TAF(II)210, TAF1 RNA polymerase II, TATA box binding protein (TBP)-associated factor, TAF1-like RNA polymerase II, TATA box binding protein (TBP)-associated factor, TAF2A2, TBP-associated factor 1-like, TBP-associated factor 210 kDa, TBP-associated factor RNA polymerase 1-like, Transcription initiation factor TFIID 210 kDa subunit, transcription initiation factor TFIID subunit 1-like | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction