missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAF5 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93025-0.1ml
This item is not returnable.
View return policy
Description
TAF5 Polyclonal antibody specifically detects TAF5 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| TAF5 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:100 | |
| TAF(II)100, TAF2DTATA box binding protein (TBP)-associated factor 2D, TAF5 RNA polymerase II, TATA box binding protein (TBP)-associated factor, TAFII100TAFII-100, TATA box binding protein (TBP)-associated factor, RNA polymerase II, D, 100kD, transcription initiation factor TFIID 100 kD subunit, Transcription initiation factor TFIID 100 kDa subunit, transcription initiation factor TFIID subunit 5 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 656-800 of human TAF5 (NP_008882.2). VLNGNCVRIFTGHKGPIHSLTFSPNGRFLATGATDGRVLLWDIGHGLMVGELKGHTDTVCSLRFSRDGEILASGSMDNTVRLWDAIKAFEDLETDDFTTATGHINLPENSQELLLGTYMTKSTPVVHLHFTRRNLVLAAGAYSPQ | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 6877 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction