missing translation for 'onlineSavingsMsg'
Learn More

Tafluprost, MedChemExpress

Product Code. 16491018
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16491018 5 mg 5mg
16458548 10 mg 10mg
16433796 25 mg 25mg
16414417 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16491018 Supplier Medchem Express Supplier No. HYB0600
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Tafluprost(AFP-168) is an anti-glaucoma prostaglandin (PG) analog.

Tafluprost(AFP-168) is an anti-glaucoma prostaglandin (PG) analog.

Specifications

Chemical Name or Material Tafluprost
CAS 209860-87-7
Color Colorless
Purity Grade Notes Research
Molecular Formula C25H34F2O5
Quantity 5 mg
Synonym AFP-168 MK2452
Solubility Information DMSO : ≥ 270 mg/mL (596.65 mM)
SMILES O=C(OC(C)C)CCC/C=C\C[C@@H]1[C@@H](/C=C/C(F)(F)COC2=CC=CC=C2)[C@H](O)C[C@@H]1O
Molecular Weight (g/mol) 452.53
Formula Weight 452.53
Percent Purity 98.0%
Grade Research
Physical Form Liquid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.