missing translation for 'onlineSavingsMsg'
Learn More

TAK-632, MedChemExpress

Product Code. 16436926
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16436926 100 mg 100mg
16402887 50 mg 50mg
16486808 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16436926 Supplier Medchem Express Supplier No. HY15767

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress TAK-632 is a potent pan-RAF inhibitor with IC50 of 1.4, 2.4 and 8.3 nM for CRAF, BRAFV600E, BRAFWT, respectively.

TAK-632 is a potent pan-RAF inhibitor with IC50 of 1.4, 2.4 and 8.3 nM for CRAF, BRAFV600E, BRAFWT, respectively.

Specifications

Chemical Name or Material TAK-632
CAS 1228591-30-7
Color White
Purity Grade Notes Research
Molecular Formula C27H18F4N4O3S
Quantity 100 mg
Solubility Information DMSO : 100 mg/mL (180.34 mM; Need ultrasonic)
SMILES O=C(NC1=CC(OC2=CC=C3N=C(NC(C4CC4)=O)SC3=C2C#N)=CC=C1F)CC5=CC=CC(C(F)(F)F)=C5
Molecular Weight (g/mol) 554.52
Formula Weight 554.52
Percent Purity 98.46%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.