missing translation for 'onlineSavingsMsg'
Learn More

TAK-733, MedChemExpress

Product Code. 16495688
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16495688 10 mg 10mg
16438387 5 mg 5mg
16421857 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16495688 Supplier Medchem Express Supplier No. HY13449
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress TAK-733 is a potent and selective MEK allosteric site inhibitor with an IC50 of 3.2 nM.

TAK-733 is a potent and selective MEK allosteric site inhibitor with an IC50 of 3.2 nM.

Specifications

Chemical Name or Material TAK-733
CAS 1035555-63-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C17H15F2IN4O4
Quantity 10 mg
Solubility Information DMSO : ≥ 33 mg/mL (65.45 mM)
SMILES O=C1C(C(NC2=CC=C(I)C=C2F)=C(F)C(N3C)=O)=C3N=CN1C[C@@H](O)CO
Molecular Weight (g/mol) 504.23
Formula Weight 504.23
Percent Purity 96.99%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.