Learn More
Invitrogen™ Talacotuzumab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA558480
 View more versions of this product
Product Code. 30284536
  • 1204.00 € / 1mg
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Talacotuzumab, also known as JNJ-56022473 (and formerly CSL-362) is a humanized IgG1 monoclonal antibody designed to bind to and neutralize CD123 (Interleukin-3 receptor alpha chain or IL3RA) with potential antineoplastic activity.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Talacotuzumab Humanized
Recombinant Monoclonal
Unconjugated
CSL362; CSL-362-AML; JNJ-56022473
Protein A
RUO
Human
Antibody
IgG1
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25mM Histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Human
1 mg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized