missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Talquetamab Humanized Recombinant Human Monoclonal Antibody
GREENER_CHOICE

Talquetamab Recombinant Monoclonal Antibody

582.00 € - 2410.00 €

Specifications

Antigen Talquetamab Humanized
Concentration 1 mg/mL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Applications ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Classification Recombinant Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30284680
GREENER_CHOICE
View Documents
Invitrogen™
MA560129
100 μg
582.00 €
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30283127
View Documents
Invitrogen™
MA560130
1 mg
2410.00 €
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Talquetamab Recombinant Monoclonal Antibody for SPR, Flow, ELISA, Functional

Talquetamab (JNJ-64407564) is a bispecific antibody that targets CD3, which is expressed on T-cells, and GPRC5D, which is highly expressed in multiple myeloma cells. Talquetamab (JNJ-64407564) is being investigated in a Phase 1 clinical study to treat relapsed or refractory multiple myeloma.
Specifications

Specifications

Talquetamab Humanized
-20°C, Avoid Freeze/Thaw Cycles
Recombinant Monoclonal
Lyophilized
IgG4
Human
JNJ-64407564
Antibody
1 mg/mL
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Unconjugated
Human
RUO
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Primary
Protein A
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.