missing translation for 'onlineSavingsMsg'
Learn More

Tamoxifen, 98%

Product Code. 15475719
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15475719 1 g 1g
15495719 500 mg 500mg
15485719 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15475719 Supplier Thermo Scientific Alfa Aesar Supplier No. J63509.03
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Tamoxifen (citofen, CAS # 10540-29-1) is an antineoplastic agent that has its effect through the estrogen receptors. | CAS: 10540-29-1 | C26H29NO | 371.52 g/mol

Inhibitor of protein kinase C and induces apoptosis in cancer cells. It is utilized in the study on lipopolysaccharide (LPS)-induced microglial activation to know its effect.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Tamoxifen is a tertiary aminobenzene that acts by competitively binding and antagonizing estrogen receptors to reduce DNA synthesis
  • When estradiol cannot bind estrogen receptors, the DNA response element is not activated
  • Tamoxifen upregulates the production of TGFb and slows tumor growth
  • Tamoxifen inhibits the production of IGF-1, a stimulator of tumor growth

Application

  • Tamoxifen may be useful in the study of tumorigenesis, growth, and inflammation
  • It is suitable for in vitro experiments
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10540-29-1
Molecular Formula C26H29NO
Molecular Weight (g/mol) 371.52
MDL Number MFCD00010454
InChI Key NKANXQFJJICGDU-QPLCGJKRSA-N
Synonym tamoxifen, trans-tamoxifen, crisafeno, soltamox, tamoxifene, diemon, tamoxifeno, tamoxifenum, citofen, istubol
PubChem CID 2733526
ChEBI CHEBI:41774
IUPAC Name (2-{4-[(1Z)-1,2-diphenylbut-1-en-1-yl]phenoxy}ethyl)dimethylamine
SMILES CC\C(=C(/C1=CC=CC=C1)C1=CC=C(OCCN(C)C)C=C1)C1=CC=CC=C1

Specifications

Color White
Quantity 1 g
Merck Index 14,9048
Solubility Information Insoluble in water; Soluble in methanol,ethanol,propanol or propylene glycol;Soluble in dimethyl sulfoxide,dichloromethane and ethanol; Insoluble in water.
Formula Weight 371.52
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Tamoxifen
compliance-icons compliance-icons
Product Identifier
  • Tamoxifen
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 1A
  • Reproductive toxicity on or via lactation
  • Reproductive toxicity Category 1A
Hazard Statement
  • H302-Harmful if swallowed.
  • H350-May cause cancer.
  • H360FD-May damage fertility. May damage the unborn child.
  • H362-May cause harm to breast-fed children.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P263-Avoid contact during pregnancy/while nursing.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Tamoxifen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.