missing translation for 'onlineSavingsMsg'
Learn More

Tamoxifen, 99.24%, MP Biomedicals™

Product Code. 11445161 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
0.25 g
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11445161 0.25 g 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11445161 Supplier MP Biomedicals Supplier No. 0215673883
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

CAS 10540-29-1
Molecular Formula C26H29NO
MDL Number MFCD00010454
Quantity 0.25 g
Synonym tamoxifen, trans-tamoxifen, crisafeno, soltamox, tamoxifene, diemon, tamoxifeno, tamoxifenum, citofen, istubol
InChI Key NKANXQFJJICGDU-QPLCGJKRSA-N
SMILES CC\C(=C(/C1=CC=CC=C1)C1=CC=C(OCCN(C)C)C=C1)C1=CC=CC=C1
IUPAC Name (2-{4-[(1Z)-1,2-diphenylbut-1-en-1-yl]phenoxy}ethyl)dimethylamine
Molecular Weight (g/mol) 371.52
PubChem CID 2733526
ChEBI CHEBI:41774
Show More Show Less
compliance-icons
Product Identifier
  • Tamoxifen, Free Base
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1A
  • Reproductive toxicity Category 1A
Hazard Statement
  • H350-May cause cancer.
  • H362-May cause harm to breast-fed children.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P263-Avoid contact during pregnancy/while nursing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: tamoxifen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.