missing translation for 'onlineSavingsMsg'
Learn More

Tamoxifen Citrate, MedChemExpress

Product Code. 16481379
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16481379 1 g 1g
16478497 5 g 5g
16421706 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16481379 Supplier Medchem Express Supplier No. HY13757
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Tamoxifen Citrate (ICI 46474) is an orally active, selective estrogen receptor modulator (SERM) which blocks estrogen action in breast cells and can activate estrogen activity in other cells, such as bone, liver, and uterine cells.Tamoxifen Citrate is a potent Hsp90 activator and enhances the Hsp90 molecular chaperone ATPase activity.

Tamoxifen Citrate also potent inhibits infectious EBOV Zaire and Marburg (MARV) with IC50 of 0.1 μM and 1.8 μM, respectively. Tamoxifen Citrate activates autophagy and induces apoptosis.

Specifications

Chemical Name or Material Tamoxifen Citrate
CAS 54965-24-1
Color White
Purity Grade Notes Research
Molecular Formula C32H37NO8
Quantity 1 g
Synonym ICI 46474(Z)-Tamoxifen Citratetrans-Tamoxifen Citrate
Solubility Information DMSO : 50 mg/mL (88.71 mM; Need ultrasonic) ∣Ethanol : 10 mg/mL (17.74 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES CC/C(C1=CC=CC=C1)=C(C2=CC=C(OCCN(C)C)C=C2)\C3=CC=CC=C3.O=C(CC(C(O)=O)(O)CC(O)=O)O
Molecular Weight (g/mol) 563.64
Formula Weight 563.64
Percent Purity 98.04%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.