missing translation for 'onlineSavingsMsg'
Learn More

Tamoxifen, MP Biomedicals

Product Code. 15712262 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15712262 5 g 5g
11455161 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15712262 Supplier MP Biomedicals Supplier No. 0215673894
  • ONLINE EXCLUSIVE PRICE
    891.00 €
    844.20 € / 5g
    Save 46.80 €
    5% Off
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Tamoxifen, White Powder

  • It has been used to facilitate the recombination of ect2flox allele in mouse organs.
  • It has also been used to study its effect on lipopolysaccharide (LPS)-induced microglial activation.
  • Protein kinase C inhibitor (IC50 = 50-200μM depending on assay conditions in MCF-7 cells or IC50 = 100μM in rat brain).
  • It is a prodrug that is metabolized to active metabolites 4-hydroxytamoxifen (4-OHT) and endoxifen by cytochrome P450 isoforms CYP2D6 and CYP3A4.

Chemical Identifiers

CAS 10540-29-1
Molecular Formula C26H29NO
Molecular Weight (g/mol) 371.52
MDL Number MFCD00010454
InChI Key NKANXQFJJICGDU-QPLCGJKRSA-N
SMILES CC\C(=C(/C1=CC=CC=C1)C1=CC=C(OCCN(C)C)C=C1)C1=CC=CC=C1

Specifications

Melting Point 96°C to 100°C
Color White
Loss on Drying 0.0021
Refractive Index KR5919600
Quantity 5 g
Synonym [Z]-1-[p-Dimethylaminoethoxyphenyl]-1,2-diphenyl-1-butene, (Z)-2-[4-(1,2-Diphenyl-1-butenyl)phenoxy]-N,N-dimethylethanamine, ICI 47699, trans-Tamoxifen
Solubility Information Practically insoluble in water (< 0.01% at 20°C); soluble in methanol, ethanol, 2-propanol, propyleneglycol, chloroform (50mg/mL - clear, colorless to faint yellow solution) or DMSO
IUPAC Name (2-{4-[(1Z)-1,2-diphenylbut-1-en-1-yl]phenoxy}ethyl)dimethylamine
Physical Form Powder
Assay 0.9935
compliance-icons compliance-icons
Product Identifier
  • Tamoxifen, Free Base
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1A
  • Reproductive toxicity Category 1A
Hazard Statement
  • H350-May cause cancer.
  • H360-May damage fertility or the unborn child.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P263-Avoid contact during pregnancy/while nursing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: tamoxifen free base
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.