missing translation for 'onlineSavingsMsg'
Learn More

TANK Monoclonal Antibody (OTI1G2), TrueMAB™, OriGene

Product Code. 17523314
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17523314 30 μL 30µL
17503314 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17523314 Supplier Origene Technologies Supplier No. TA810317S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TANK Monoclonal antibody specifically detects TANK in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen TANK
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1G2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene KLF12
Gene Accession No. Q9Y4X4
Gene Alias C86182; E430026L09Rik; ITRAF; I-TRAF; LOW QUALITY PROTEIN: TRAF family member-associated NF-kappa-B activator; OTTHUMP00000200694; OTTHUMP00000200695; OTTHUMP00000200696; OTTHUMP00000200698; OTTHUMP00000200700; OTTHUMP00000200703; OTTHUMP00000200704; TANK; TANK-like protein; TRAF family member associated NFKB activator; TRAF family member-associated Nf-kappa B activator; TRAF family member-associated NF-kappa-B activator; TRAF family member-associated NFKB activator; TRAF interacting protein TANK; TRAF2; TRAF-interacting protein; zgc:153048
Host Species Mouse
Immunogen Full length human recombinant protein of human KLF12 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11278
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.