missing translation for 'onlineSavingsMsg'
Learn More

Tannic acid, MedChemExpress

Product Code. 16451148
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16451148 5 g 5g
16432379 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16451148 Supplier Medchem Express Supplier No. HYB2136
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Tannic acid is a novel hERG channel blocker with IC50 of 3.4 μM.

Tannic acid is a novel hERG channel blocker with IC50 of 3.4 μM.

Specifications

Chemical Name or Material Tannic acid
CAS 1401-55-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C76H52O46
Quantity 5 g
Solubility Information DMSO : 100 mg/mL (58.78 mM; Need ultrasonic) ∣H2O : ≥ 100 mg/mL (58.78 mM)
SMILES O=C(C1=CC(OC(C2=CC(O)=C(O)C(O)=C2)=O)=C(O)C(O)=C1)OC3[C@H](OC(C4=CC(OC(C5=CC(O)=C(O)C(O)=C5)=O)=C(O)C(O)=C4)=O)[C@@H](OC(C6=CC(OC(C7=CC(O)=C(O)C(O)=C7)=O)=C(O)C(O)=C6)=O)[C@H](OC(C8=CC(OC(C9=CC(O)=C(O)C(O)=C9)=O)=C(O)C(O)=C8)=O)[C@@H](COC(C%10=CC(OC(C%11=CC(O)=C(O)C(O)=C%11)=O)=C(O)C(O)=C%10)=O)O3
Molecular Weight (g/mol) 1701.2
Formula Weight 1701.2
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.