Learn More
Invitrogen™ TAP2 Recombinant Rabbit Monoclonal Antibody (5I10B3)

Rabbit Recombinant Monoclonal Antibody
176.00 € - 436.00 €
Specifications
| Antigen | TAP2 |
|---|---|
| Clone | 5I10B3 |
| Concentration | 0.8 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA, Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30283361
|
Invitrogen™
MA558169 |
20 μL |
176.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30282449
![]() |
Invitrogen™
MA558170 |
100 μL |
436.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Immunogen sequence: QPNLPSPGTL APTTLQGVVK FQDVSFAYPN RPDRPVLKGL TFTLRPGEVT ALVGPNGSGK STVAALLQNL YQPTGGQVLL DEKPISQYEH CYLHSQVVSV GQEPVLFSGS VRNNIAYGLQ SCEDDKVMAA AQAAHADDFI QEMEHGIYTD VGEKGSQLAA GQKQRLAIAR ALVRDPRVLI LDEATSALDV QCEQALQDWN SRGDRTVLVI AHRLQTVQRA HQILVLQEGK LQKLAQL.
The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MDR/TAP subfamily. Members of the MDR/TAP subfamily are involved in multidrug resistance. This gene is located 7 kb telomeric to gene family member ABCB2. The protein encoded by this gene is involved in antigen presentation. This protein forms a heterodimer with ABCB2 in order to transport peptides from the cytoplasm to the endoplasmic reticulum. Mutations in this gene may be associated with ankylosing spondylitis, insulin-dependent diabetes mellitus, and celiac disease. Alternative splicing of this gene produces two products which differ in peptide selectivity and level of restoration of surface expression of MHC class I molecules.Specifications
| TAP2 | |
| 0.8 mg/mL | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| Q03519 | |
| 6891 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 450-686 of human TAP2 (NP_001276972.1). | |
| Antibody |
| 5I10B3 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| TAP2 | |
| ABC transporter, MHC 2; ABC18; ABCB3; AI462429; Antigen peptide transporter 2; APT2; ATP dependent transport protein family member; ATP-binding cassette sub-family B member 3; ATP-binding cassette, sub-family B (MDR/TAP), member 3; Cim; D6S217E; Ham2; Ham-2; Histocompatibility antigen modifier 2; jas; MTP2; Peptide supply factor 2; peptide transporter involved in antigen processing 2; Peptide transporter PSF2; Peptide transporter TAP2; PSF2; PSF-2; Really interesting new gene 11 protein; RING11; TAP2; Tap-2; Transporter 2 ABC (ATP binding cassette); transporter 2 ATP-binding cassette sub-family B; transporter 2, ABC (ATP binding cassette); transporter 2, ATP binding cassette subfamily B member; transporter 2, ATP-binding cassette, sub-family B (MDR/TAP); Y1 | |
| TAP2 | |
| Primary | |
| Protein A |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.