missing translation for 'onlineSavingsMsg'
Learn More

Taprenepag isopropyl, MedChemExpress

Product Code. 16478008
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16478008 10 mg 10mg
16410478 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16478008 Supplier Medchem Express Supplier No. HY19998
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Taprenepag isopropyl is a highly selective EP2 receptor agonist.

Taprenepag isopropyl is a highly selective EP2 receptor agonist.

Specifications

Chemical Name or Material Taprenepag isopropyl
CAS 1005549-94-9
Color White
Purity Grade Notes Research
Molecular Formula C27H28N4O5S
Quantity 10 mg
Synonym PF-04217329
Solubility Information DMSO : 200 mg/mL (384.17 mM; Need ultrasonic)
SMILES O=S(C1=CN=CC=C1)(N(CC2=CC(OCC(OC(C)C)=O)=CC=C2)CC3=CC=C(N4C=CC=N4)C=C3)=O
Molecular Weight (g/mol) 520.6
Formula Weight 520.6
Percent Purity 98.77%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.