missing translation for 'onlineSavingsMsg'
Learn More

TAPS sodium salt, 98%

Product Code. 15413819
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15413819 100 g 100g
15403819 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15413819 Supplier Thermo Scientific Alfa Aesar Supplier No. J61189.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Zwitterionic buffer | CAS: 91000-53-2 | C7H16NNaO6S | 265.256 g/mol

TAPS sodium salt is used in biochemistry and molecular biology. It has a useful pH range of 7.7 - 9.1. It has been utilized in capillary electrophoresis of such materials as DNA and DNA-dye complexes. Dyes have been separated by planar chromatography with electro osmotic flow with 1 mM TAPS in the mobile phase. The stability constants of metal ions with TAPS have been investigated by capillary electrophoresis. It has also been shown to inhibit connexin channel activity. The activity of various mutants of carbonic anhydrase I and carbonic anhydrase II has been studied in a variety of buffers, including TAPS.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
TAPS sodium salt is used in biochemistry and molecular biology. It has a useful pH range of 7.7 - 9.1. It has been utilized in capillary electrophoresis of such materials as DNA and DNA-dye complexes. Dyes have been separated by planar chromatography with electro osmotic flow with 1 mM TAPS in the mobile phase. The stability constants of metal ions with TAPS have been investigated by capillary electrophoresis. It has also been shown to inhibit connexin channel activity. The activity of various mutants of carbonic anhydrase I and carbonic anhydrase II has been studied in a variety of buffers, including TAPS.

Solubility
Soluble in water (333.33 mg/ml).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91000-53-2
Molecular Formula C7H16NNaO6S
Molecular Weight (g/mol) 265.256
MDL Number MFCD00069903
InChI Key FEGYIWVHCSRXCG-UHFFFAOYSA-M
PubChem CID 23667519
IUPAC Name sodium;3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]propane-1-sulfonate
SMILES C(CNC(CO)(CO)CO)CS(=O)(=O)[O-].[Na+]

Specifications

Color White
Physical Form Powder
Quantity 100 g
Linear Formula (HOCH2)3CNH(CH2)3SO3Na
Solubility Information Soluble in water (333.33mg/ml).
Formula Weight 265.26
Percent Purity 98%
Chemical Name or Material TAPS sodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.