missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAS2R38 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-95168-0.02ml
This item is not returnable.
View return policy
Description
TAS2R38 Polyclonal antibody specifically detects TAS2R38 in Rat samples. It is validated for Western Blot
Specifications
| TAS2R38 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| PTCphenylthiocarbamide tasting, T2R38, T2R61PTC bitter taste receptor, taste receptor type 2 member 38, Taste receptor type 2 member 61, taste receptor, type 2, member 38 | |
| A synthetic peptide corresponding to a sequence within amino acids 150-250 of human TAS2R38 (NP_789787.4). CSCICTVLCVWCFFSRPHFTVTTVLFMNNNTRLNWQIKDLNLFYSFLFCYLWSVPPFLLFLVSSGMLTVSLGRHMRTMKVYTRNSRDPSLEAHIKALKSLV | |
| 0.02 mL | |
| GPCR | |
| 5726 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction