Learn More
TAT Antibody (OTI3G6), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-44843
Product Code. 30306166
  • 735.00 € / 100µg
Description
TAT Monoclonal antibody specifically detects TAT in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TAT
Monoclonal
Unconjugated
Lyophilized powder ( 1X PBS, pH 7.3,)
Mouse
>95%
RUO
Primary
For reconstitution, we recommend adding 100uL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiment, we strongly recommend performing another round of desalting process.
Store at -20 degrees C. Avoid freeze/thaw cycles.
Lyophilized
Western Blot
OTI3G6
Western Blot 1:2000
EC 2.6.1.5, L-tyrosine:2-oxoglutarate aminotransferase, tyrosine aminotransferase, tyrosine aminotransferase, cytosolic
Full length human recombiant protein of human TAT (NP_000344) produced in HEK293T cell.
100 μg
Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction
6898
Human, Mouse, Rat
Antibody
IgG2a