missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Tat-Beclin 1 D11 Autophagy Inducing Peptide - Retroinverso form

Product Code. 18694148 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
1 mg
5 mg
Unit Size:
0.10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18694148 1 mg 1mg
18699676 5 mg 5mg
18614158 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18694148 Supplier Novus Biologicals™ Supplier No. NBP249888

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Peptide - Retroinverso form

TRUSTED_SUSTAINABILITY

Specifications

Species Non-species Specific
Content And Storage Store at -20°C in powder form. Store at -80°C once reconstituted.
Gene Alias Autophagy Inducing peptide, Tat-Beclin, Tat-Beclin 1, Tat-Beclin 1 peptide, Tat-Beclin peptide, Tat-D11
Molecular Weight (g/mol) MolecularWeight-theroretical: 3.08 kDa
Quantity 1 mg
Research Category Autophagy
Storage Buffer This product is supplied lyophilized. Purity is ≥ to 97% (HPLC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.