Learn More
TAZ/WWTR1 Antibody (CL0371) [DyLight 550], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-52926R
Product Code. 30289865
  • 737.00 € / 0.10mL
Description
TAZ/WWTR1 Monoclonal antibody specifically detects TAZ/WWTR1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TAZ/WWTR1
Monoclonal
DyLight 550
FLJ27004, FLJ45718, TAZDKFZp586I1419, transcriptional co-activator with PDZ-binding motif, Transcriptional coactivator with PDZ-binding motif, WW domain containing transcription regulator 1, WW domain-containing transcription regulator protein 1
This antibody was developed using a recombinant protein derived from Q9GZV5, with the exact immunogen sequence remaining proprietary.
0.1 mL
Chromatin Research, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Markers, Transcription Factors and Regulators
25937
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown, Immunocytochemistry/Immunofluorescence
CL0371
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified