missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tazobactam sodium salt

Product Code. 15545146 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15545146 1 g 1g
15555146 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15545146 Supplier Thermo Scientific Chemicals Supplier No. 455370010
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89785-84-2
Molecular Formula C10H11N4NaO5S
Molecular Weight (g/mol) 322.27
InChI Key RFMIKMMOLPNEDG-LEFOPYBQSA-M
IUPAC Name sodium (2S,3S)-3-methyl-4,4,7-trioxo-3-[(1H-1,2,3-triazol-1-yl)methyl]-4λ⁶-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
SMILES [Na+].C[C@]1(CN2C=CN=N2)[C@@H](N2C(CC2=O)S1(=O)=O)C([O-])=O

Specifications

Type Tazobactam Sodium Salt
Melting Point 140.0°C to 147.0°C
Quantity 1 g
Solubility Information Solubility aq. soln.: soluble.
Formula Weight 322.27
Chemical Name or Material Tazobactam sodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.