missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBC1D1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-82928
This item is not returnable.
View return policy
Description
TBC1D1 Polyclonal specifically detects TBC1D1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| TBC1D1 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| KIAA1108TBC, TBC1, TBC1 (tre-2/USP6, BUB2, cdc16) domain family, member 1, TBC1 domain family member 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| TBC1D1 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:ENDLLNKRLKLDYEEITPCLKEVTTVWEKMLSTPGRSKIKFDMEKMHSAVGQGVPRHHRGEIWKFLAEQFHLKHQFPSKQQPKD | |
| 0.1 mL | |
| Signal Transduction | |
| 23216 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction