missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBC1D24 Mouse, anti-Human, Clone: 1C7C4, Proteintech
Mouse Monoclonal Antibody
145.00 € - 410.00 €
Specifications
| Antigen | TBC1D24 |
|---|---|
| Clone | 1C7C4 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, ELISA |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18045585
|
Proteintech
68566-1-IG-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18017055
|
Proteintech
68566-1-IG-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The TBC1D24 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TBC1D24. This antibody recognizes human, rat, mouse antigen. The TBC1D24 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| TBC1D24 | |
| 1000 μg/mL | |
| Liquid | |
| Human, Rat, Mouse | |
| KIAA1171, TBC1 domain family member 24, TBC1 domain family, member 24, TBC1D24 | |
| Primary |
| 1C7C4 | |
| Western Blot, ELISA | |
| Mouse | |
| 57465 | |
| IgG1 | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title