missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBE Buffer Mix (Dry Powder Mix of Tris/Boric Acid/EDTA/Electrophoresis), Fisher BioReagents
Description
Electrophoresis buffer.
Specifications
Specifications
| Chemical Name or Material | TBE Buffer Mix |
| CAS | 77-86-1, 6381-92-6, 10043-35-3 |
| Color | White |
| pH | 8.2 to 8.4 |
| Physical Form | Solid |
| Quantity | 86.1 g |
| DNase | DNase free |
| Packaging | Amber glass bottle |
| Synonym | Tris-Boric Acid-EDTA Buffer Mix |
| Grade | Electrophoresis |
Safety and Handling
Product Identifier
- Tris borate EDTA 10X powder
- Danger
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H360FD-May damage fertility. May damage the unborn child.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?