Learn More
TBK1 Antibody (S01-1G3), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-19537-50ul
 View more versions of this product
Product Code. 18683195
  • 421.00 € / 50µL
Description
TBK1 Monoclonal antibody specifically detects TBK1 in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TBK1
Monoclonal
0.3 mg/mL
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Rabbit
Affinity purified
RUO
Primary
Human, Mouse, Rat
Purified
Western Blot
S01-1G3
Unconjugated
EC 2.7.11, EC 2.7.11.1, FLJ11330, NAK serine/threonine-protein kinase TBK 1, NAKserine/threonine-protein kinase TBK1, NF-kappa-B-activating kinase, T2K, TANK-binding kinase 1NF-kB-activating kinase, TBK 1
A synthetic peptide of human TBK1 (Uniprot # Q9UHD2)
50 μg
Phospho Specific, Protein Kinase, Signal Transduction
29110
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.