missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This gene encodes a member of the beta-transducin protein family. Most proteins of the beta-transducin family are involved in regulatory functions. This protein is possibly involved in some intracellular signaling pathway. This gene is deleted in Williams-Beuren syndrome, a developmental disorder caused by deletion of multiple genes at 7q11.23.
Specifications
Specifications
| Antigen | TBL2 |
| Applications | Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.31 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gene | TBL2 |
| Gene Accession No. | Q9R099, Q9Y4P3 |
| Gene Alias | TBL2, transducin (beta) like 2, Transducin beta like protein 2, WBSCR13, WS betaTRP |
| Gene Symbols | Tbl2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?