missing translation for 'onlineSavingsMsg'
Learn More

TBS, Tris Buffered Saline, 10X Solution, pH 7.4, Molecular Biology, Fisher BioReagents™

Product Code. 10153103
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
Packaging:
Nalgene Poly Bottle
Poly Bottle
Unit Size:
1 L
100mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10153103 500 mL Poly Bottle 500mL
10648973 100 mL Poly Bottle 100mL
10776834 1 L Nalgene Poly Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10153103 Supplier Fisher BioReagents Supplier No. BP2471500
Spotlight Offers
Use promo code 28036
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
This item is not returnable. View return policy

TBS, Tris Buffered Saline, 10X Solution, pH 7.4, Molecular Biology

  • Save time and simplify your buffer preparation step by using concentrated Fisher BioReagents 10X TBS buffer solution - simply dilute as needed
  • 10X solution contains 1.37M NaCl, 0.027M KCl and 0.25M tris
  • To prepare a 1X solution, mix one volume of Fisher BioReagents 10X TBS solution with 9 volumes of ultrapure water, such as BP2819
  • Filtered and autoclaved - ready to use stock TBS buffer
  • Tested for the absence of DNase, RNase, and protease
  • TBS buffer is often used in immunodetection techniques such as Western blotting protocols
  • Intended for laboratory research use only
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tris Buffered Saline
Name Note 10X Solution
CAS 7447-40-7, 7647-01-0, 7647-14-5, 77-86-1, 7732-18-5
Boiling Point 100°C
Color Colorless
pH 7.5
Physical Form Liquid
Quantity 500 mL
DNase DNase free
Purity Grade Notes DNase-, RNase- and Protease-Free
Packaging Poly Bottle
Protease Protease free
Synonym TBS
Grade Molecular Biology
Concentration or Composition (by Analyte or Components) 10X solution contains 1.37M Sodium Chloride, 0.027M Potassium Chloride, and 0.25M Tris/Tris-HCl.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.