missing translation for 'onlineSavingsMsg'
Learn More

TBS, Tris Buffered Saline, 1X Solution, pH 7.4, Molecular Biology, Fisher BioReagents

Product Code. 10467583
Change view
Click to view available options
Quantity:
1 L
Packaging:
Nalgene Poly Bottle
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10467583 1 L Nalgene Poly Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10467583 Supplier Fisher BioReagents Supplier No. BP24721
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
  • Save time and simplify your buffer preparation step by using ready-made Fisher BioReagents 1X TBS buffer solution
  • Eliminates the hassle of dilution or waiting for powder to dissolve
  • Filtered and autoclaved - ready to use TBS buffer
  • Tested for the absence of DNase, RNase, and protease
  • TBS buffer is often used in immunodetection techniques such as Western blotting protocols
  • Intended for laboratory research use only
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tris Buffered Saline
Name Note 1X Solution, pH 7.4
CAS 7647-14-5, 7447-40-7, 7647-01-0, 7732-18-5, 77-86-1
Boiling Point 100°C
Color Colorless
pH 7.4
Physical Form Liquid
Quantity 1 L
DNase DNase free
Packaging Nalgene Poly Bottle
Protease Protease free
Synonym TBS
Grade Molecular Biology
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.