missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBX3 Rabbit anti-Mouse, Rat, Clone: 3G9F1, Novus Biologicals™
Rabbit Monoclonal Antibody
190.00 € - 496.00 €
Specifications
| Antigen | TBX3 |
|---|---|
| Clone | 3G9F1 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18311597
|
Novus Biologicals
NBP3-16848-20UL |
20 μg |
190.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18355822
|
Bio-Techne
NBP3-16848-100UL |
100 μg |
496.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TBX3 Monoclonal antibody specifically detects TBX3 in Mouse, Rat samples. It is validated for Western BlotSpecifications
| TBX3 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis, Apoptosis, Cardiovascular Biology, Cell Biology | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 6926 | |
| IgG | |
| Affinity purified |
| 3G9F1 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse, Rat | |
| bladder cancer related protein XHL, T-box 3, T-box protein 3, T-box transcription factor TBX3, TBX3-ISO, ulnar mammary syndrome, UMS, XHL | |
| A synthetic peptide corresponding to a sequence within amino acids 644-743 of human TBX3 (O15119). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title