missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCEB1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94131-0.02ml
This item is not returnable.
View return policy
Description
TCEB1 Polyclonal antibody specifically detects TCEB1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| TCEB1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| eloC, Elongin 15 kDa subunit, elongin-C, RNA polymerase II transcription factor SIII subunit C, SIII, SIII p15, transcription elongation factor B (SIII), polypeptide 1 (15kD, elongin C), transcription elongation factor B (SIII), polypeptide 1 (15kDa, elongin C), transcription elongation factor B polypeptide 1, transcription elongation factor B, polypeptide 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-112 of human TCEB1 (NP_001191791.1). MDGEEKTYGGCEGPDAMYVKLISSDGHEFIVKREHALTSGTIKAMLSGPGQFAENETNEVNFREIPSHVLSKVCMYFTYKVRYTNSSTEIPEFPIAPEIALELLMAANFLDC | |
| 0.02 mL | |
| Stem Cell Markers | |
| 6921 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction