Learn More
Invitrogen™ TCEB2 Recombinant Rabbit Monoclonal Antibody (23GB5615), Invitrogen™
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA553914
 View more versions of this product
Product Code. 30089079
  • 562.00 € / 100µL
Description
SIII, also known as elongin, is a general transcription elongation factor that increases the RNA polymerase II transcription elongation past template-encoded arresting sites. Subunit A is transcriptionally active and its transcription activity is strongly enhanced by binding to the dimeric complex of the SIII regulatory subunits B and C (elongin BC complex).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TCEB2
Recombinant Monoclonal
Unconjugated
ELOB
0610040H15Rik; 18 kDa, elongin B; cb162; ELOB; Elongin 18 kDa subunit; elongin B; elongin, 18-kD subunit; elongin-B; hypothetical protein LOC617277; RNA polymerase II transcription factor SIII p18 subunit; RNA polymerase II transcription factor SIII subunit B; SIII; SIII p18; TCEB 2; Tceb2; tceb2 protein; transcription elongation factor B (SIII) polypeptide 2 (18kD, elongin B); transcription elongation factor B (SIII), polypeptide 2; transcription elongation factor B (SIII), polypeptide 2 (18 kDa, elongin B); transcription elongation factor B (SIII), polypeptide 2 (18kD, elongin B); transcription elongation factor B (SIII), polypeptide 2 (18kDa, elongin B); transcription elongation factor B polypeptide 2; transcription elongation factor B polypeptide 2; elongin-B; transcription elongation factor B subunit 2; wu:fb47e08
Rabbit
Affinity chromatography
RUO
6923
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
23GB5615
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q15370
ELOB
A synthesized peptide derived from human TCEB2 (1-50AA).