missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCF-2/HNF-1 beta Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 6 publications
Brand: Novus Biologicals NBP1-89680-25ul
This item is not returnable.
View return policy
Description
TCF-2/HNF-1 beta Polyclonal specifically detects TCF-2/HNF-1 beta in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Chromatin Immunoprecipitation (ChIP).
Specifications
| TCF-2/HNF-1 beta | |
| Polyclonal | |
| Western Blot 0.04-0.4 ug/ml, Simple Western 1:30, Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:1000 - 1:2500, Chromatin Immunoprecipitation (ChIP) | |
| P35680 | |
| HNF1B | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:KEVLVQALEELLPSPNFGVKLETLPLSPGSGAEPDTKPVFHTLTNGHAKGRLSGDEGSEDGDDYDTPPILKELQALNTEEAAEQRAEVDRMLSEDPWRAAKMIKGYMQQH | |
| 25 μL | |
| Diabetes Research | |
| 6928 | |
| Human, Mouse | |
| IgG |
| ChIP Assay, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| FJHN, hepatocyte nuclear factor 1-beta, HNF1 beta A, HNF1 homeobox B, HNF-1B, HNF1beta, HNF-1-beta, HNF2, Homeoprotein LFB3, LFB3, LF-B3, MODY5HPC11, TCF-2, TCF2transcription factor 2, hepatic; LF-B3; variant hepatic nuclear factor, Transcription factor 2, transcription factor 2, hepatic, Variant hepatic nuclear factor 1, vHNF1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction