missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCF19 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93718-0.02ml
This item is not returnable.
View return policy
Description
TCF19 Polyclonal antibody specifically detects TCF19 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| TCF19 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| SC1(TCF19)-6, SC1-1, SC1SC1(TCF19)-7, SCI(TCF19)-4, TCF-19, transcription factor 19, Transcription factor SC1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-290 of human TCF19 (NP_009040.2). MLPCFQLLRIGGGRGGDLYTFHPPAGAGCTYRLGHRADLCDVALRPQQEPGLISGIHAELHAEPRGDDWRVSLEDHSSQGTLVNNVRLPRGHRLELSDGDLLTFGPEGPPGTSPSEFYFMFQQVRVKPQDFAAITIPRSRGEARVGAGFRPMLPSQGAPQRPLSTFSPAPKATLILNSIGSLSKLRPQPLTFSPSWGGPKSLPVPAPPGEMGTTPSAPPQRNRRKSVHRVLAELDDESEPPENPPPVLMEPRKKLRVDKAPLTPTGNRRGRPRKYPVSAPMAPPAVGGGE | |
| 0.02 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 6941 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction