missing translation for 'onlineSavingsMsg'
Learn More

TCF21 Antibody (SR1424), Novus Biologicals™

Rabbit Monoclonal Antibody

349.00 € - 472.00 €

Specifications

Antigen TCF21
Clone SR1424
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:80, Immunoprecipitation 1:60
Applications Western Blot, Flow Cytometry, Immunoprecipitation
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18686414
View Documents
Novus Biologicals
NBP3-22128-100ul
100 μg
472.00 €
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18601394
View Documents
Novus Biologicals
NBP3-22128-50ul
50 μg
349.00 €
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TCF21 Monoclonal antibody specifically detects TCF21 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation
Specifications

Specifications

TCF21
Western Blot 1:500-1:2000, Flow Cytometry 1:80, Immunoprecipitation 1:60
Monoclonal
Purified
RUO
Human
bHLHa23BHLHA23, Capsulin, Class A basic helix-loop-helix protein 23, epicardin, Pod-1, POD1pod-1, Podocyte-expressed 1, TCF-21, transcription factor 21
A synthesized peptide derived from human TCF21 (Uniprot #: O43680)
Primary
Store at -20° C. Avoid freeze/thaw cycles.
SR1424
Western Blot, Flow Cytometry, Immunoprecipitation
Unconjugated
Rabbit
Cancer, Stem Cell Markers
PBS, pH 7.4, 150mM NaCl, 50% glycerol.
6943
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.