missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TCHH Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121557
This item is not returnable.
View return policy
Description
This protein is an intermediate filament-associated protein that associates in regular arrays with keratin intermediate filaments (KIF) of the inner root sheath cells of the hair follicle and the granular layer of the epidermis. It later becomes cross-linked to KIF by isodipeptide bonds. It may serve as scaffold protein, together with involucrin, in the organization of the cell envelope or even anchor the cell envelope to the KIF network.
Specifications
| TCHH | |
| Polyclonal | |
| Unconjugated | |
| TCHH | |
| AHF; AI597080; hair follicle protein AHF; Tchh; THH; THL; TRHY; Trichohyalin | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 310588, 7062 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q07283 | |
| TCHH | |
| Phe1851-Pro1943 (Accession Q07283), with N-terminal His Tag. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction