missing translation for 'onlineSavingsMsg'
Learn More

TCLP Base/Neutral/Acid Extractable Spike Solution, SPEX CertiPrep™

Product Code. 15286326 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15286326 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15286326 Supplier SPEX Certiprep Supplier No. TCLPBNA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
1,4-Dichlorobenzene106-46-72000μg/mL
2,4,5-Trichlorophenol95-95-42000μg/mL
2,4,6-Trichlorophenol88-06-22000μg/mL
2,4-Dinitrotoluene121-14-22000μg/mL
2-Methylphenol95-48-72000μg/mL
3-Methylphenol108-39-42000μg/mL
4-Methylphenol106-44-52000μg/mL
Hexachlorobenzene118-74-12000μg/mL
Hexachlorobutadiene87-68-32000μg/mL
Hexachloroethane67-72-12000μg/mL
Nitrobenzene98-95-32000μg/mL
Pentachlorophenol87-86-52000μg/mL
Pyridine110-86-12000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material TCLP Base/Neutral/Acid Extractable Spike Solution
Packaging 2mL amber prescored glass ampule
CAS 106-46-7, 95-95-4, 88-06-2, 121-14-2, 95-48-7, 108-39-4
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
Analyte or Component Names 1,4-Dichlorobenzene,2
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Element 1,4-Dichlorobenzene
Solvent or Matrix MeCl
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 2000μg/mL

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.