missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TCP-1 epsilon Monoclonal Antibody (PK/29/23/8d)

Description
In Western blot, this antibody detects a band at ~60 kD in lysates of heat-shocked Hela cells. Rat anti CCT epsilon antibody, clone PK/29/23/8d recognizes the epsilon polypeptide of the CCT chaperonin molecule complex.
This gene encodes a molecular chaperone that is member of the chaperonin containing TCP1 complex (CCT), also known as the TCP1 ring complex (TRiC). This complex consists of two identical stacked rings, each containing eight different proteins. Unfolded polypeptides enter the central cavity of the complex and are folded in an ATP-dependent manner. The complex folds various proteins, including actin and tubulin. Alternate transcriptional splice variants of this gene have been observed but have not been thoroughly characterized.
Specifications
Specifications
| Antigen | TCP-1 epsilon |
| Applications | Gel Shift, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | PK/29/23/8d |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide |
| Gene | CCT5 |
| Gene Accession No. | P48643, P80316, Q68FQ0 |
| Gene Alias | CCT5; CCTE; CCT-epsilon; chaperonin containing TCP1 subunit 5; chaperonin containing Tcp1, subunit 5 (epsilon); chaperonin subunit 5 (epsilon); epididymis secretory protein Li 69; HEL-S-69; Kiaa0098; mKIAA0098; PNAS-102; T-complex protein 1 subunit epsilon; TCP-1 epsilon; TCP-1-epsilon; TCPE; unm tf212; unm_tf212; wu:fb54h08 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?