Learn More
Invitrogen™ TCP-1 epsilon Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA522093
Description
Recommended positive controls: 293T, Molt-4, Mouse brain. Predicted reactivity: Mouse (94%), Rat (94%), Zebrafish (83%), Xenopus laevis (86%), Pig (96%), Chicken (88%), Rhesus Monkey (100%), Chimpanzee (100%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a molecular chaperone that is member of the chaperonin containing TCP1 complex (CCT), also known as the TCP1 ring complex (TRiC). This complex consists of two identical stacked rings, each containing eight different proteins. Unfolded polypeptides enter the central cavity of the complex and are folded in an ATP-dependent manner. The complex folds various proteins, including actin and tubulin. Alternate transcriptional splice variants of this gene have been observed but have not been thoroughly characterized.
Specifications
| TCP-1 epsilon | |
| Polyclonal | |
| Unconjugated | |
| CCT5 | |
| CCT5; CCTE; CCT-epsilon; chaperonin containing TCP1 subunit 5; chaperonin containing Tcp1, subunit 5 (epsilon); chaperonin subunit 5 (epsilon); epididymis secretory protein Li 69; HEL-S-69; Kiaa0098; mKIAA0098; PNAS-102; T-complex protein 1 subunit epsilon; TCP-1 epsilon; TCP-1-epsilon; TCPE; unm tf212; unm_tf212; wu:fb54h08 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 12465, 22948 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| P48643, P80316 | |
| CCT5 | |
| Recombinant protein encompassing a sequence within the center region of human TCP1 epsilon. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.