missing translation for 'onlineSavingsMsg'
Learn More

TCP1 alpha Antibody (91a), Novus Biologicals™

Rat Monoclonal Antibody

Brand:  Novus Biologicals NBP1-97505-0.025MG

 View more versions of this product

Product Code. 18066627

  • 193.00 € / 25µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TCP1 alpha Monoclonal specifically detects TCP1 alpha in Human, Mouse, Rat, Porcine, Bovine, Canine, C. elegans, Drosophila, Guinea Pig, Plant, Primate, Rabbit, Yeast samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TCP1 alpha
Monoclonal
Unconjugated
PBS and 50% Glycerol with 0.09% Sodium Azide
TCP1
Recombinant mouse TCP-1alpha (C-terminal fragment).
0.025 mg
Cancer, Hypoxia, Membrane Trafficking and Chaperones, Stem Cell Markers
6950
Store at -20C. Avoid freeze-thaw cycles.
IgG2a
Flow Cytometry, Immunoprecipitation, Western Blot, Immunocytochemistry, Immunohistochemistry, Immunofluorescence
91a
Western Blot 1:1000, Flow Cytometry 1:100, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
CCT1T-complex protein 1 subunit alpha, CCTa, CCT-alpha, D6S230E, tailless complex polypeptide 1, t-complex 1, T-complex protein 1, alpha subunit, TCP-1-alpha
Rat
Protein G purified
RUO
Primary
Yeast, Mouse, Guinea Pig, Rabbit, Primate, Human, Canine, Pig, C. elegans, Rat, Bovine, Drosophila, Plant
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only